Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Interchangeable blocks for block heaters, digital, SB1
Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, 1,5 ml tubes, 24
Catalog # JENC357-126
Supplier:   JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Catalog # JENC357-126
Supplier:  JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, 1,5 ml tubes, 24
Price
271,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
Specifications
  • No. of Holes:
    24
  • Description:
    Interchangeable block
  • Seq:
    05
  • For:
    1,5 ml tubes