Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, 2 ml tubes, 20
Catalog # JENC357-124
Supplier: JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Catalog # JENC357-124
Supplier: JENCONS SCIENTIFIC
Jump to:
Product Details & Documents
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
Specifications
- No. of Holes:20
- Description:Interchangeable block
- Seq:03
- For:2 ml tubes