Interchangeable blocks for block heaters, digital, SB1
Interchangeable plain block, without holes, -, —
Catalog # JENC357-122
Supplier: JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Catalog # JENC357-122
Supplier: JENCONS SCIENTIFIC
Jump to:
Product Details & Documents
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
Specifications
- No. of Holes:—
- Description:Interchangeable plain block, without holes
- Seq:01
- For:-