Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Diphosphorus pentoxide, Sigma-Aldrich®

Diphosphorus pentoxide, Sigma-Aldrich®

Glass bottle for solids, 250 g
Catalog # SIAL298220-250G
Corrosive
Supplier:   Merck
Diphosphorus pentoxide, Sigma-Aldrich®
Catalog # SIAL298220-250G
Supplier:  Merck
CAS Number:  1314-56-3
Diphosphorus pentoxide, Sigma-Aldrich®

Glass bottle for solids, 250 g

CAS Number:  1314-56-3
Price
40,60 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Phosphorus(V) oxide, di-Phosphorus pentoxide, Phosphorus pentoxide, Phosphoric anhydride
  • CAS Number:1314-56-3
  • Formula:P₂O₅
  • MDL Number:MFCD00011440
  • Molecular Weight:141.94 g/mol
  • Merck Index:13,07441
  • Melting Pt:562 °C
  • Density:2.253 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-236-1
  • UN:1807
  • ADR:8,II
  •  :M-Clarity™ quality level = MQ200
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.