Sampling swabs, Dryswab™
Product Details & Documents
These dry swabs are ideal for collecting specimens or samples for analysis or for transferring materials during a diagnostic procedure.
- Sterile, ready-to-use
- Available with different shaft and head materials
- Rayon swabs are particularly suitable for the recovery of microorganisms
Labelled tubes: Each swab comes self-contained and ready to use with its own tube for easy transport to the laboratory. The swab is mounted in the tube plug which serves as a convenient holder so there is no need for direct handling of the swab. Each tube and cap assembly is sealed with a tamper evident label for assurance of sterility and which includes the batch details for traceability. The caps are colour coded according to the type of swab.
Peel pouches: Individual or multiple swabs are packed sterile and ready to use within an easy-to-open tamper evident peel pouch. These are especially suitable where the specimen is to be tested immediately or transferred to another transport container perhaps as part of a test kit. Each peel pouch has product and batch details printed for reference and traceability.
Bulk packed: Swabs are packed loose in autoclavable paper bags of 100.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
- Description:Dry swabs, standard tip
- Seq:06
- Shaft/head:Wood/cotton