MRS Broth modified, Vegitone 1 * 500 g
Catalog # SIAL38944-500G-F
Supplier: Merck
Frequently Bought Together
Hydrochloric acid 0.02 mol/l (0.02 N), AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
n-Hexane
Supplier: Roth Carl
(±)-2-Ethyl-1-hexanol 99%
Supplier: Thermo Fisher Scientific
p-Xylene 99% for spectroscopy
Supplier: Thermo Fisher Scientific
n-Pentane 99+%, extra pure
Supplier: Thermo Fisher Scientific
Dibutyl ether, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
Chlorobenzene ≥99%
Supplier: Thermo Fisher Scientific
(±)-2-Methyltetrahydrofuran 99+% stabilised, pure
Supplier: Thermo Fisher Scientific
Tetrabutylammonium hydroxide 0.4 M in water for HPLC
Supplier: Thermo Fisher Scientific
Tetrahydrofuran 99.8% unstabilised for HPLC
Supplier: Thermo Fisher Scientific