Bromine 99+%, extra pure
Glass bottle, plastic-coated, 500 mL
Supplier: Thermo Fisher Scientific
Bromine 99+%, extra pure
Catalog # ACRO402845000
Supplier: Thermo Fisher Scientific
CAS Number: 7726-95-6
Jump to:
Product Details & Documents
- CAS Number:7726-95-6
- Formula:Br₂
- MDL Number:MFCD00010896
- Molecular Weight:159.81 g/mol
- Merck Index:13,01378
- Boiling Pt:58.7 °C (1013 hPa)
- Melting Pt:–7.2 °C
- Density:3.111 g/cm³
- Storage Temperature:Ambient
- EINECS:231-778-1
- UN:1744
- ADR:8,I
Documents
Specifications
Specification Test Results
- Appearance:Red-brown Liquid
- Titration Iodometric:≥99.0 %
- Chlorine (Cl):≤500 ppm
- Iodine (I):≤10 ppm
- Heavy metals (as Pb):≤2 ppm
Specifications
- Pack type:Glass bottle, plastic-coated
- Pk:500 mL



