Determines consistency of almost any viscous substance by measuring the distance a material flows under its own weight during a given time interval.Can be used by food, paint, chemical, and other industries to pre-determine formulas for standardizing 1 * 1 items
Catalog # VWRMNP23270-004
Supplier: VWR Products
Frequently Bought Together