HEAD PRINT FOR SARTORIUS DATA PRINTER YDP03-0CE1 1 * 1 items
SARB69Y03137
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Xylene (mixture of isomers) ≥98%, TECHNICAL
: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
: VWR Chemicals