Ammonium acetate ≥97%
Plastic bottle, 1.000 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 631-61-8 Molecular Formula: C2H7NO2 Formula Weight: 77.08 Flash Point: 136°(277°F) Odour: Ammonia-like Boiling Point: dec. Beilstein Reference: 4186741 Merck Reference: 14,495 MDL No.: MFCD00013066
Synonyms: Acetic acid ammonium salt
- CAS Number:631-61-8
- Formula:H₃CCOONH₄
- MDL Number:MFCD00013066
- Molecular Weight:77.08 g/mol
- Merck Index:14,495
- Boiling Pt:Dec.
- Melting Pt:∼114 °C (dec.)
- Density:1.17
- Flash Pt:136 °C (277 °F)
- Storage Temperature:Ambient
- EINECS:211-162-9
Specifications
Specification Test Results
- Form:Crystals or powder or crystalline powder
- Identification (FTIR):Conforms
- Ammonia as ammonium acetate:≥96.0% to ≤104.0%
- Water Content (Karl Fischer Titration):≤5.0%
- Appearance (Color):White
Specifications
- Pack type:Plastic bottle
- Pk:1.000 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



