Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Ethylhexanal ≥97%

2-Ethylhexanal ≥97%

Glass bottle for solids, 250 g
Catalog # B23471.30
Flammable
Irritant
2-Ethylhexanal ≥97%
Catalog # B23471.30
CAS Number:  123-05-7
2-Ethylhexanal ≥97%
2-Ethylhexanal ≥97%

Glass bottle for solids, 250 g

CAS Number:  123-05-7
Price
94,10 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 97% CAS No.: 123-05-7 Molecular Formula: C8H16O Formula Weight: 128.21 Flash Point: 50°(122°F) Refractive Index: 1.4155 Melting Point: -100° Boiling Point: 163° Beilstein Reference: 1700556 UN No.: 1989 MDL No.: MFCD00006987

  • CAS Number:123-05-7
  • Formula:C₈H₁₆O
  • MDL Number:MFCD00006987
  • Molecular Weight:128.21 g/mol
  • Boiling Pt:163 °C
  • Melting Pt:< –100 °C
  • Density:0.819
  • Flash Pt:50 °C (122 °F)
  • Storage Temperature:Ambient
  • EINECS:204-596-5
  • UN:1989
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.