Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Ethylhexanal ≥97%
Glass bottle for solids, 50 g
  B23471.18
2-Ethylhexanal ≥97%
  B23471.18
 :  123-05-7
2-Ethylhexanal ≥97%
2-Ethylhexanal ≥97%
Glass bottle for solids, 50 g
 :  123-05-7
37,90 €
1 * 50 g
Purity: 97% CAS No.: 123-05-7 Molecular Formula: C8H16O Formula Weight: 128.21 Flash Point: 50°(122°F) Refractive Index: 1.4155 Melting Point: -100° Boiling Point: 163° Beilstein Reference: 1700556 UN No.: 1989 MDL No.: MFCD00006987
  •  :123-05-7
  •  :C₈H₁₆O
  •  :MFCD00006987
  •  :128.21 g/mol
  •  :163 °C
  •  :< –100 °C
  •  :0.819
  •  :50 °C (122 °F)
  •  :Ambient
  •  :204-596-5
  •  :1989
  •  :3,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g