Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Colony counters
Colony counter manual with magnifying glass ø 120 mm with lighting and support
  710-0108
Colony counters
  710-0108
 :  
Colony counters
Colony counters
Colony counter manual with magnifying glass ø 120 mm with lighting and support
Choice of models with a fibre marker with a built-in switch and /or a metal-tipped pen that are attached to the base. The marker supplied with the counter allows marked cells to be added to the screen one by one.
  • Ø 100 or 120 mm biconvex lamp
  • X2 magnification
220 V voltage, 50 Hz, 0,2 A.
  • Description:
    Colony counter manual with magnifying glass ø 120 mm with lighting and support
  • Seq:
    02
Accessories
  •  
    Description
  • 710-0111
    Centering Device 100 mm
    1 * 1 items - 61,00 €
  •  
    Description
  • 710-0110
    Centering Device 80 mm
    1 * 1 items - 61,00 €
  •  
    Description
  • 710-0109
    Centering Device 60 mm
    1 * 1 items - 61,00 €