TICKOPUR R 32 - 5 LITRES, SPECIAL CLEANER, WITHOUT COMPLEXING AGENTS, WITH CORROSION PROTECTION, CONCENTRATE, GENTLE TO MATERIAL, DO SAGE 0.25 - 5 %, MILDLY ALKALINE, PH 9.9 (1 %) 1 * 5 L
Catalog # BAND832
Supplier: BANDELIN-ELECTRONIC
Frequently Bought Together