Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Replacement Cables
Replacement Cables
Replacement cable. Connect an Agilent 5890 GC to remote-start another piece of equipment or to start the Agilent 5890 GC from that piece of equipment, 1,8 m
Catalog # REST20657
Supplier:   Restek
Replacement Cables
Catalog # REST20657
Supplier:  Restek
Replacement Cables
Replacement Cables
Replacement cable. Connect an Agilent 5890 GC to remote-start another piece of equipment or to start the Agilent 5890 GC from that piece of equipment, 1,8 m
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
These are replacement cables for Agilent GCs, integrators and autosamplers.
Delivery information: Instructions and writing diagrams included.
Specifications
  • Description:
    Replacement cable. Connect an Agilent 5890 GC to remote-start another piece of equipment or to start the Agilent 5890 GC from that piece of equipment
  • Length:
    1,8 m
  • Seq:
    02