Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Buffer solutionpH 7,00±0,02 (20 °C) 1 * 1 L
Catalog # ROTHA518.1
Supplier:
Roth Carl
Expand image
Buffer solutionpH 7,00±0,02 (20 °C) 1 * 1 L
Catalog # ROTHA518.1
Supplier:
Roth Carl
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Sodium hydroxide 0.25 mol/l (0.25 N) in aqueous solution
Supplier: Roth Carl