Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Wipes for delicate tasks, KIMTECH SCIENCE*

Wipes for delicate tasks, KIMTECH SCIENCE*

White, 200×205 mm, 200 tissues/box
Catalog # ROTHP502.1
Supplier:   Roth Carl
Wipes for delicate tasks, KIMTECH SCIENCE*
Catalog # ROTHP502.1
Supplier:  Roth Carl
Wipes for delicate tasks, KIMTECH SCIENCE*

White, 200×205 mm, 200 tissues/box

Price
8,20 €
Log in for availability
1 * 200 items
Product Details & Documents

These tissues are made of 100 % pure pulp. They are excellent for wiping smaller amounts of liquid and for cleaning pipettes and slides.

  • Two-ply, white, bleached without using chlorine
  • Particularly absorbent and soft
  • Suitable for application with solvents and chemicals
  • Low-fluffing and especially ideal for sensitive areas
Specifications
  • Packed:
    200 tissues/box
  • Colour:
    White
  • L×W:
    200×205 mm
  • Seq:
    02