Order Entry
Slovenia
Orders LinkContactUsLinkComponent
VWR®, Laboratory Jacks

VWR®, Laboratory Jacks

122×150 mm, 80 kg, 80 - 278 mm, Yellow
Catalog # 245-0007
Supplier:   VWR Collection
VWR®, Laboratory Jacks
Catalog # 245-0007
Supplier:  VWR Collection
VWR®, Laboratory Jacks
VWR®, Laboratory Jacks

122×150 mm, 80 kg, 80 - 278 mm, Yellow

Price
492,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Anodised aluminium. With M10 threaded hole in the middle of the top plate for attaching retort rods (starting from item number 245-0006).

  • Smooth adjustment of working height
  • Extreme stability
  • Exceptionally sturdy
  • Corrosion resistant and non susceptible to a wide range of chemicals
Specifications
  • Max. static load capacity:
    80 kg
  • Plate size:
    122×150 mm
  • Working height:
    80 - 278 mm
  • Colour:
    Yellow
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.