Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Separating funnels, pear shaped, Quickfit®
Separating funnels, pear shaped, Quickfit®
500 ml, 19/26 NS, 24/29 NS
Catalog # 532-0086
Supplier:   SciLabware
Separating funnels, pear shaped, Quickfit®
Catalog # 532-0086
Supplier:  SciLabware
Separating funnels, pear shaped, Quickfit®
Separating funnels, pear shaped, Quickfit®
500 ml, 19/26 NS, 24/29 NS
Price
140,00 €
Log in for availability
1 * 1 items
Product Details & Documents
PYREX®, borosilicate glass.
  • Without graduation
  • Excellent chemical resistance
  • Excellent temperature resistance
Funnel stem with cone to allow easy connection to Quickfit® flask with interchangeable PTFE key stopcock.
Certifications: Complies with ISO 4800 type 2.
Specifications
  • Cone size:
    19/26 NS
  • Socket size:
    24/29 NS
  • Capacity:
    500 ml
  • Seq:
    08