4-Cyclohexylaniline
Glass bottle for solids, 1 g
Catalog # MAYBSB00566.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:6373-50-8
- Formula:C₁₂H₁₇N
- MDL Number:MFCD00001454
- Molecular Weight:175.27 g/mol
- Boiling Pt:166 °C (13 mmHg)
- Melting Pt:55 °C
- Density:1,13 g/cm³ (–100.15 °C)
- Storage Temperature:Ambient
- EINECS:228-918-9
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



