4-Cyclohexylaniline
Glass bottle for solids, 250 mg
Catalog # MAYBSB00566.250
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:6373-50-8
- Formula:C₁₂H₁₇N
- MDL Number:MFCD00001454
- Molecular Weight:175.27 g/mol
- Boiling Pt:166 °C (13 mmHg)
- Melting Pt:55 °C
- Density:1,13 g/cm³ (–100.15 °C)
- Storage Temperature:Ambient
- EINECS:228-918-9
- UN:2811
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



