4-Nitroindole
Glass bottle for solids, 250 mg
Catalog # MAYBRH01152.250
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 4-Nitro-1H-indole
- CAS Number:4769-97-5
- Formula:C₈H₆N₂O₂
- MDL Number:MFCD00010056
- Molecular Weight:162.15 g/mol
- Melting Pt:206…209 °C
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



