Bis(2-iodophenyl)disulphide
Glass bottle for solids, 1 g
Catalog # MAYBNRB00619.1
Supplier: Thermo Scientific
Bis(2-iodophenyl)disulphide
Catalog # MAYBNRB00619.1
Supplier: Thermo Scientific
CAS Number: 87797-73-7
Jump to:
Product Details & Documents
- CAS Number:87797-73-7
- Formula:C₁₂H₈IS₂
- MDL Number:MFCD00099178
- Molecular Weight:343.23 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



