4-Hydroxyindole
Glass bottle for solids, 250 mg
Catalog # CHMPBTB07067.250
Supplier: CHEMPUR
Jump to:
Product Details & Documents
Synonyms: Indol-4-ol, 4-Indolol, 4-Hydroxy-1H-indole, 1H-Indol-4-ol
- CAS Number:2380-94-1
- Formula:C₈H₇NO
- MDL Number:MFCD00005667
- Molecular Weight:133.15 g/mol
- Boiling Pt:100 °C (0,01 mmHg)
- Boiling Pt:100 °C (0.01 mmHg)
- Melting Pt:102…104 °C
- EINECS:219-177-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



