Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Pharmacist's spatulas
Pharmacist's spatulas
Hardened blade, 150 mm, 18/22 mm
Catalog # 231-0554
Supplier:   RSG
Pharmacist's spatulas
Catalog # 231-0554
Supplier:  RSG
Pharmacist's spatulas
Pharmacist's spatulas
Hardened blade, 150 mm, 18/22 mm
Price
131,00 €
Log in for availability
1 * 10 items
Product Details & Documents
Stainless steel blade, wooden handle.
  • Available with flexible blade (Remanit 4310) or with hardened blade (INOX 4034)
  • 75 to 150 mm: conical blade
Specifications
  • Description:
    Hardened blade
  • Blade width:
    18/22 mm
  • Blade length:
    150 mm