Sample vessel
Catalog # HACHLZQ035_U
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Sample vessels 25 mL, polystyrol, 200 pieces 1 * 200 items Catalog #HACHLZQ035
Product HACHLZQ035 is a replacement for HACHLZQ035_U but varies by Brand, Packaging or Quantity.
Product Details & Documents
Sample vessel made from PS. For aqueous solutions, acids and bases.
For use with autosampler LPV122/LZQ031.