Order Entry
Slovenia
ContactUsLinkComponent
Tube Microfilterfuge with microfiltration membrane cellulose acetate 0,45µm 1 * 50 items
Catalog # RAIN7016-025
Supplier:  RAININ
Tube Microfilterfuge with microfiltration membrane cellulose acetate 0,45µm 1 * 50 items
Catalog # RAIN7016-025
Supplier:  RAININ
Frequently Bought Together