Electrode cleaning solutions
Pepsin-HCl solution, for cleaning junctions with protein contamination, treatment time about 1 h, 6×250 ml, 6
Catalog # 662-0573
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Cleaning agent Pepsin/Hydrochloric acid Plastic bottle 500 ML 1 * 500 mL Catalog #83603.260
Product 83603.260 is similar to 662-0573 but may have a different manufacturer and/or packaging.
PEP/PH,PEPSIN CLEANING SOLUTION ONLY FOR ELECTRODES WITH LIQUID ELECTROLYTES3 BOTTLES, 250 ML EACH 1 * 3 items Catalog #662-8291
Product 662-8291 is similar to 662-0573 but may have a different manufacturer and/or packaging.
Cleaning agent Pepsin/Hydrochloric acid Plastic bottle 1L 1 * 1 L Catalog #83603.290
See more alternativesProduct 83603.290 is similar to 662-0573 but may have a different manufacturer and/or packaging.
Product Details & Documents
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:6
- Description:Pepsin-HCl solution, for cleaning junctions with protein contamination, treatment time about 1 h, 6×250 ml



