Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Counting chambers, BLAUBRAND® Thoma

Counting chambers, BLAUBRAND® Thoma

Counting chamber, with clips, double ruling, Thoma, 0,1 mm
Catalog # BRND718020
Supplier:   Brand
Counting chambers, BLAUBRAND® Thoma
Catalog # BRND718020
Supplier:  Brand
Counting chambers, BLAUBRAND® Thoma
Counting chambers, BLAUBRAND® Thoma

Counting chamber, with clips, double ruling, Thoma, 0,1 mm

Price
85,90 €
Log in for availability
1 * 1 items
Product Details & Documents

Double ruling. The net division corresponds to the large square lying in the middle of the Neubauer chamber. The area of the small square is 0,0025 mm². Since the outer large squares are not included, the Thoma system is only used to count thrombocytes and erythrocytes.

Specifications
  • Depth of chamber:
    0,1 mm
  • Type:
    Thoma
  • Description:
    Counting chamber, with clips, double ruling
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.