Order Entry
Slovenia
ContactUsLinkComponent
Container pp-purpose 7l,general purpose containers - polypropylene (pp)+130°c,autoclavable, suitable forsterilization325x265x150mm, 9.25 l. 1 * 1 items
Catalog # 216-7328
Supplier:  SEMADENI
Container pp-purpose 7l,general purpose containers - polypropylene (pp)+130°c,autoclavable, suitable forsterilization325x265x150mm, 9.25 l. 1 * 1 items
Catalog # 216-7328
Supplier:  SEMADENI
Frequently Bought Together