Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Stators, Rheodyne®
Stators, Rheodyne®
7060-065 stainless steel stator for 7060L
Catalog # 554-2649
Supplier:   IDEX RHEODYNE
Stators, Rheodyne®
Catalog # 554-2649
Supplier:  IDEX RHEODYNE
Stators, Rheodyne®
Stators, Rheodyne®
7060-065 stainless steel stator for 7060L
Price
966,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Stators are available in 316 stainless steel, PEEK and proprietary materials. Rheodyne® stator materials have been researched and selected for their physical and mechanical strengths. Stators only need replacing if the ports or sealing surfaces become damaged.
DuraLife® stators are stainless steel stators with the wear surface coated to prolong valve life.
DuraLife® II stators are titanium base metal with coated wear surfaces.
Specifications
  • Description:
    7060-065 stainless steel stator for 7060L
  • Seq:
    07