Restriction Endonucleases Dpn I (New England BioLabs 176S) 1 * 1.000 EU
Catalog # NEWER0176S
Supplier: NEW ENGLAND BIOLABS
Restriction Endonucleases Dpn I (New England BioLabs 176S) 1 * 1.000 EU
Catalog # NEWER0176S
Supplier: NEW ENGLAND BIOLABS
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Decontamination solution, RNase AWAY®
Supplier: Thermo Fisher Scientific
Peqlab peqGOLD, Agarose, Universal
Supplier: VWR Chemicals
VWR®, Petri Dish, PS
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® Standard Line Bottles, Narrow Neck, Round, PET, with Screw Caps
Supplier: VWR Collection
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
Proteinase K solution
Supplier: PanReac AppliChem
Tissue DNA kit, genomic DNA isolation from tissue, E.Z.N.A.®
Supplier: OMEGA BIO-TEK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Decontamination solution, DNA AWAY®
Supplier: Thermo Fisher Scientific
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Centrifugal filtration units, Vivaspin® 20
Supplier: Sartorius
Safe-Lock Tubes, 1,5 ml - Microtubes
Supplier: EPPENDORF
KLD Enzyme Mix - 25 rxns 1 * 25 AU
Supplier: NEW ENGLAND BIOLABS