Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

1,2,6-Hexanetriol 97+%, extra pure

Glass bottle for solids, 500 g
Catalog # 139835000.
1,2,6-Hexanetriol 97+%, extra pure
Catalog # 139835000.
CAS Number:  106-69-4
1,2,6-Hexanetriol 97+%, extra pure
1,2,6-Hexanetriol 97+%, extra pure

Glass bottle for solids, 500 g

CAS Number:  106-69-4
Price
521,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  1,2,6-Trihydroxyhexane
  • CAS Number:106-69-4
  • Formula:C₆H₁₄O₃
  • MDL Number:MFCD00002976
  • Molecular Weight:134.18 g/mol
  • Boiling Pt:178 °C (6.7 mmHg)
  • Melting Pt:25…32 °C
  • Density:1.109 g/cm³
  • Flash Pt:198 °C
  • Storage Temperature:Ambient
  • EINECS:203-424-6

 

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Viscous liquid (May solidify at about 32°C)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4750 to 1.4770 (20°C, 589 nm)
  • Water:
    ≤0.2 % (K.F.)
  • GC:
    ≥97.0 %
  • Appearance of solution:
    (10% in ethanol) clear colorless to
  • pH:
    4.0 to 7.0 (30 % at 20°C)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications