1,2,6-Hexanetriol 97+%, extra pure
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
1,2,6-Hexanetriol 97+%, extra pure
Catalog # 139831000.
Supplier: Thermo Fisher Scientific
CAS Number: 106-69-4
Jump to:
Product Details & Documents
Synonyms: 1,2,6-Trihydroxyhexane
- CAS Number:106-69-4
- Formula:C₆H₁₄O₃
- MDL Number:MFCD00002976
- Molecular Weight:134.18 g/mol
- Boiling Pt:178 °C (6.7 mmHg)
- Melting Pt:25…32 °C
- Density:1.109 g/cm³
- Flash Pt:198 °C
- Storage Temperature:Ambient
- EINECS:203-424-6
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Viscous liquid (May solidify at about 32°C)
- Infrared spectrum:Conforms
- Refractive index:1.4750 to 1.4770 (20°C, 589 nm)
- Water:≤0.2 % (K.F.)
- GC:≥97.0 %
- Appearance of solution:(10% in ethanol) clear colorless to
- pH:4.0 to 7.0 (30 % at 20°C)
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



