2-Ethylhexanal 96%
Glass bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:123-05-7
- Formula:C₈H₁₆O
- MDL Number:MFCD00006987
- Molecular Weight:128.21 g/mol
- Boiling Pt:163 °C (1013 hPa)
- Melting Pt:–90 °C
- Density:0.8195 g/cm³
- Flash Pt:50 °C
- Storage Temperature:Ambient
- EINECS:204-596-5
- UN:1191
- ADR:3,III
Documents
ACRO41009
SDS
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4140 to 1.4180 (20°C, 589 nm)
- GC:≥95 %
- Specific gravity:(20/20°C) 0.819 to 0.824
- Acid value:≤8 mg KOH/g
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



