Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Fluorocyclohexanone 96%

2-Fluorocyclohexanone 96%

Bottle
Catalog # APOSPC201281-5G
New Product
Supplier:   Apollo Scientific
2-Fluorocyclohexanone 96%
Catalog # APOSPC201281-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  694-82-6
2-Fluorocyclohexanone 96%

Bottle

CAS Number:  694-82-6
Price
870,00 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:694-82-6
  • Formula:C₆H₉FO
  • MDL Number:MFCD01076476
  • Molecular Weight:116.14 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications