Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Fluorocyclohexanone 96%

2-Fluorocyclohexanone 96%

Bottle
Catalog # APOSPC201281-1G
New Product
Supplier:   Apollo Scientific
2-Fluorocyclohexanone 96%
Catalog # APOSPC201281-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  694-82-6
2-Fluorocyclohexanone 96%

Bottle

CAS Number:  694-82-6
Price
183,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:694-82-6
  • Formula:C₆H₉FO
  • MDL Number:MFCD01076476
  • Molecular Weight:116.14 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.