FMOC-3-(4-PYRIDYL)-L-ALANINE 1 * 250 items
Catalog # MSPPA-0333-250G
Supplier: MARKET SOURCE PART PROCESS
New Product
FMOC-3-(4-PYRIDYL)-L-ALANINE 1 * 250 items
Catalog # MSPPA-0333-250G
Supplier: MARKET SOURCE PART PROCESS
New Product
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetrachloroethylene ≥99.9%, SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
n-Pentane ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Tetrachloroethylene ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
2,2,4-Trimethylpentane ≥99.5%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Tetrahydrofuran 99.5% stabilised for analysis
Supplier: Thermo Fisher Scientific