Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SENSISWAB KIT- 10 X 5 TESTS 1 * 1 KIT
Catalog # MSPPHU0030102
Supplier:
MARKET SOURCE PART PROCESS
New Product
Expand image
SENSISWAB KIT- 10 X 5 TESTS 1 * 1 KIT
Catalog # MSPPHU0030102
Supplier:
MARKET SOURCE PART PROCESS
New Product
Frequently Bought Together
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Sodium hydroxide 32% in aqueous solution, AnalaR NORMAPUR® analytical reagent, for nitrogen analysis
Supplier: VWR Chemicals
Sodium hydroxide 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Test strips, nitrate/nitrite, Dosatest
Supplier: VWR Chemicals