Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Polystyrene Standard Reference for MIR-ATR Technique
Polystyrene standard reference material for measurement by MIR-ATR technique
Catalog # BRUK1867838
New Product
Supplier:   BRUKER DALTRONIK
Polystyrene Standard Reference for MIR-ATR Technique
Catalog # BRUK1867838
Supplier:  BRUKER DALTRONIK
New Product
CAS Number:  
Polystyrene Standard Reference for MIR-ATR Technique
Polystyrene standard reference material for measurement by MIR-ATR technique
CAS Number:  
Price
649,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Polystyrene standard reference material 'BRM 1921' for measurement by ATR technique including certificate for validation of the wavenumber calibration of the spectrometer in the mid-infrared spectral range mounted in a sample card.
Specifications
Specifications
  • Description:
    Polystyrene standard reference material for measurement by MIR-ATR technique