Order Entry
Slovenia
ContactUsLinkComponent
G-Biosciences TBE Running Buffer (10X)
G-Biosciences TBE Running Buffer (10X)
Catalog # R025
Supplier:  G-Biosciences
CAS Number:  
G-Biosciences TBE Running Buffer (10X)
Catalog # R025
Supplier:  G-Biosciences
CAS Number:  
Specifications
  • Pk:
    1 L
  • Description:
    TBE (10X)
Specifications
About this item
The primary use of TBE running buffer is in agarose electrophoresis for the separation of nucleic acids (DNA and RNA).
  • 10X concentrated solution: 0,9 M tris-borate, 0,01 M EDTA (pH 8,3)
The buffer maintains consistent pH and conductivity, enabling sharp band resolution and protecting nucleic acids from degradation during electrophoresis.