GOODJOB® Black Grip Gloves
Supplier: FELDTMANN HELMUT
New Product
Certificates
About this item
Black grip 100% polyester, red gloves.
Seamless knitted, 13 G
Comfortable wearing comfort
Good fit
Coating: latex, crinkled, black
Certifications:
EN 388.
Packaging:
Packing: 120 pairs.
Specifications
- Catalog No:
- FELD0519/9
- FELD0519/8
- FELD0519/7
- FELD0519/11
- FELD0519/10
- Material:
- Polyester
- Polyester
- Polyester
- Polyester
- Polyester