Human eNOS ELISA Kit
Human eNOS ELISA kit, 96 tests
Catalog # ABCAAB263878-1X96
Supplier: Abcam
Jump to:
Product Details & Documents
Human eNOS ELISA kit is a single-wash 90-min Simplestep used to quantify Human eNOS with a sensitivity of 77 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.
Specifications
Specifications
- Description:Human eNOS ELISA kit
- Tests per kit:96 tests
- Primary antibody reactivity:Human
- Sample type:Cell lysate
- Detection range:234,38 - 15000 pg/ml
- Storage temperature:4 °C
- Assay Type:Sandwich
- Time to Results:1 h 30 min
- Cat. No.:ABCAAB263878-1X96
- Assay duration:One step
- Detection method:Colorimetric
- Sensitivity:77 pg/ml
- Format:Pre-coated
- Target protein:eNOS
- Regulatory status:RUO
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



