Order Entry
Slovenia
Orders LinkContactUsLinkComponent
Mouse Tweak ELISA Kit

Mouse Tweak ELISA Kit

Mouse Tweak ELISA Kit, 96 tests
Catalog # ABCAAB113320-96
Supplier:   Abcam
Mouse Tweak ELISA Kit
Catalog # ABCAAB113320-96
Supplier:  Abcam
CAS Number:  
Mouse Tweak ELISA Kit
Mouse Tweak ELISA Kit

Mouse Tweak ELISA Kit, 96 tests

CAS Number:  
Price
1 250,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Mouse Tweak ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Tweak in Mouse in plasma, cell culture supernatant, serum samples.

Specifications

Specifications

  • Description:
    Mouse Tweak ELISA Kit
  • Tests per kit:
    96 tests
  • Cat. No.:
    ABCAAB113320-96
  • Assay duration:
    Multiple steps
  • Detection method:
    Colorimetric
  • Sensitivity:
    0,3 pg/ml
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Sample type:
    Plasma, Cell culture supernatant, Serum
  • Target protein:
    Tweak
  • Detection range:
    0,27 - 200 pg/ml
  • Storage temperature:
    4 °C
  • Regulatory status:
    RUO
  • Assay Type:
    Sandwich

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.