Human CES2 ELISA Kit
Human CES2 ELISA kit, 96 tests
ABCAAB313529-96TST
: Abcam
Human CES2 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human CES2 in Human in cell culture supernatant, plasma, serum samples.
- Description:Human CES2 ELISA kit
- Tests per kit:96 tests
- Cat. No.:ABCAAB313529-96TST
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:1,6 ng/ml
- Format:Pre-coated
- Primary antibody reactivity:Human
- Sample type:Cell culture supernatant, Plasma, Serum
- Target protein:CES2
- Detection range:1,6 - 400 ng/ml
- Storage temperature:4 ℃
- Regulatory status:RUO
- Assay Type:Sandwich



