Order Entry
Slovenia
ContactUsLinkComponent
Pre-Coated 96 Well Microplate (12×8 well strips) SimpleStep ELISA®
Pre-Coated 96 Well Microplate (12×8 well strips) SimpleStep ELISA®
Catalog # ABCAAB206978-1
Supplier:  Abcam
CAS Number:  
undefined
Pre-Coated 96 Well Microplate (12×8 well strips) SimpleStep ELISA®
Catalog # ABCAAB206978-1
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Pre-Coated 96 Well Microplate (12 x 8 well strips) SimpleStep ELISA®
  • Storage temperature:
    4 °C
  • Tests per kit:
    1 unit

Specifications

About this item

Pre-Coated 96 Well Microplate (12×8 well strips) SimpleStep ELISA® is to use for the development of your own SimpleStep ELISA kit - use with SimpleStep ELISA Custom ELISA kit (ab270552).