Anti-c-Fos (phospho S32) Rabbit Monoclonal Antibody [clone: cFosS32-BA9]
Supplier: Abcam
Certificates
About this item
Rabbit monoclonal [cFosS32-BA9] to c-Fos (phospho S32).
Type: Primary
Antigen: c-Fos
Clonality: Monoclonal
Clone: cFosS32-BA9
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Frequently Bought Together