- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:RAGE
- Description:Mouse RAGE ELISA kit (small sample volume)
- Sample type:Serum, plasma, and other biological fluids
- Cross reactivity:No significant cross-reactivity or interference between mouse RAGE and analogues was observed.
- Detection method:Colorimetric
- Time to Results:3 h 30 min
- Shelf life:Store for 6 months at 4 °C
- Detection range:0,78 - 50 ng/ml
- Storage temperature:4 °C
- Sensitivity:0,47 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Mouse RAGE ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse RAGE in serum, plasma, and other biological fluids. This ELISA kit only requires 25 µl of sample volume(s).
- Small Sample Volume: Requires as little as 25 µl of sample!
- Assay Precision: Intra-Assay: CV <10%, Inter-Assay: CV <10%
Mouse RAGE ELISA kit employs the sandwich Enzyme-Linked Immunoassay technique for the quantitative measurement of mouse RAGE in serum, plasma, and other biological fluids. An antibody specific for RAGE has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the RAGE present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-RAGE Antibody, which binds the captured RAGE present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Avidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of RAGE captured in each well. The concentration of RAGE can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.