Human Recombinant Furin (from HEK293 Cells)
Supplier: Antibodies.com
New Product
Certificates
About this item
Partial length Human Furin, expressed in HEK293 cells.
Caution: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.
Specifications
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:HEK293 cells
- Species:Human
- Storage conditions:Shipped at 4 °C. Lyophilised: Store at −20...−80 °C. Reconstituted: Aliquot and store at −80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Gene ID:UniprotID# P09958
- Reconstitution instructions:Reconstitute with distilled sterile water.
- Carrier-free:N
- Protein synonyms:Dibasic-processing enzyme|FUR|PACE|Paired basic amino acid residue-cleaving enzyme|PCSK3
- Protein/peptide name:Furin
- Purity:>85% by SDS-PAGE and Coomassie blue staining
- Molecular weight:The protein has a predicted molecular mass of 61,0 kDa after removal of the signal peptide. The apparent molecular mass of FURIN-His is approximately 55 - 70 kDa due to glycosylation.
- Sequence:FURIN (Gln27-Ala574) + 6×His Tag
- Concentration:Reconstitution dependent
- Formulation:Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation.
- Shipping temperature:Shipped on blue ice at 4 °C
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together