
Dimethyl-2,5-pyridinedicarboxylate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:881-86-7
:
Formula:C₉H₉NO₄
MDL Number:MFCD00034767
:195.17 g/mol
Melting Pt:160…163 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together